4-Brom-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Bromopyridin-2-yl)morpholine, 4-(4-Bromo-2-pyridyl)morpholine

PY-1241-5G PY-1241-1G
COBBPY-1241-5GEA 1250 EUR
COBBPY-1241-5G COBBPY-1241-1G
4-Brom-2-(4-morpholino)pyridin
4-Brom-2-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
Molecular Weight: 243.1004 g/mol
MDL: MFCD13195616
CAS-Nummer: 1040377-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR