3-Brom-5-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(5-Bromo-3-pyridyl)morpholine, 3-Bromo-5-morpholinopyridine

PY-1103-5G PY-1103-1G
COBBPY-1103-5GEA 0 EUR
COBBPY-1103-5G COBBPY-1103-1G
3-Brom-5-(4-morpholino)pyridin
3-Brom-5-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
MDL: MFCD11656285
CAS-Nummer: 200064-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR