6-Morpholino-3-pyridinamin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine
H50699.03
H50699.MD
H50699.06
H50699.03EA
193
EUR
H50699.03
H50699.MD
H50699.06
6-Morpholino-3-pyridinamin 97%
6-Morpholino-3-pyridinamin
Purity: 97%
CAS No.: 52023-68-4
Molecular Formula: C9H13N3O
Formula Weight: 179.22
Melting Point: 127-130°
MDL No.: MFCD00119562
Formel:
C₉H₁₃N₃O Molecular Weight: 179,22 g/mol Schmelzpunkt: 127…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00119562 CAS-Nummer: 52023-68-4 EINECS: 257-613-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...