6-Morpholino-3-pyridinamin
Lieferant: BIONET RESEARCH
Synonyme:
6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine
11R-0626.500
11R-0626.1
11R-0626.5
BIOT11R-0626.500EA
0
EUR
BIOT11R-0626.500
BIOT11R-0626.1
BIOT11R-0626.5
6-Morpholino-3-pyridinamin
6-Morpholino-3-pyridinamin
Formel:
C₉H₁₃N₃O Molecular Weight: 179,22 g/mol |
MDL:
MFCD00119562 CAS-Nummer: 52023-68-4 EINECS: 257-613-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...