2-Brom-3',5'-bis(trifluormethyl)acetophenon
Lieferant: MENAI
Synonyme:
1-(3,5-Bis(trifluoromethyl)phenyl)-2-bromoethanone, 3,5-Bis(trifluoromethyl)phenacyl bromide
E2695.10
MENAE2695.10EA
0
EUR
MENAE2695.10
2-Brom-3',5'-bis(trifluormethyl)acetophenon
2-Brom-3',5'-bis(trifluormethyl)acetophenon
Formel:
C₁₀H₅BrF₆O Molecular Weight: 335.0405 g/mol |
MDL:
MFCD00792434 CAS-Nummer: 131805-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...