2'-Hydroxy-4',5'-dimethylacetophenon ≥98%

Lieferant: Thermo Scientific

Synonyme: 1-(2-Hydroxy-4,5-dimethylphenyl)ethan-1-one

H50685.03 H50685.06
H50685.03EA 23 EUR
H50685.03 H50685.06
2'-Hydroxy-4',5'-dimethylacetophenon ≥98%
2'-Hydroxy-4',5'-dimethylacetophenon

Purity: 98% CAS No.: 36436-65-4 Molecular Formula: C10H12O2 Formula Weight: 164.21 Melting Point: 88-90° MDL No.: MFCD00002306

Formel: C₁₀H₁₂O₂
Molecular Weight: 164,21 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002306
CAS-Nummer: 36436-65-4
EINECS: 253-035-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR