2'-Hydroxy-4',5'-dimethylacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(2-Hydroxy-4,5-dimethylphenyl)ethan-1-one

FL12077.POR 012077-25G
CHMPFL12077.POREA 0 EUR
CHMPFL12077.POR FLUO012077-25G
2'-Hydroxy-4',5'-dimethylacetophenon
2'-Hydroxy-4',5'-dimethylacetophenon
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.21 g/mol
MDL: MFCD00002306
CAS-Nummer: 36436-65-4
EINECS: 253-035-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR