3',5'-Di-tert-butyl-4'-hydroxyacetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12722.06
A12722.14
A12722.06EA
85.6
EUR
A12722.06
A12722.14
3',5'-Di-tert-butyl-4'-hydroxyacetophenon ≥98%
3',5'-Di-tert-butyl-4'-hydroxyacetophenon
Purity: 98%
CAS No.: 14035-33-7
Molecular Formula: C16H24O2
Formula Weight: 248.37
Melting Point: 146-147°
MDL No.: MFCD00017520
Formel:
C₁₆H₂₄O₂ Schmelzpunkt: 146…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017520 CAS-Nummer: 14035-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...