3',5'-Dimethyl-4'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

546402-25G 546402-5G
SIAL546402-25GEA 350 EUR
SIAL546402-25G SIAL546402-5G
3',5'-Dimethyl-4'-hydroxyacetophenon, Sigma-Aldrich®
3',5'-Dimethyl-4'-hydroxyacetophenon
Formel: C₁₀H₁₂O₂
MDL: MFCD00195574
CAS-Nummer: 5325-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR