3',5'-Dimethyl-2'-hydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 2-Acetyl-4,6-dimethylphenol, 2'-Hydroxy-3',5'-dimethylacetophenone

FL12076.POR 012076-25G
CHMPFL12076.POREA 0 EUR
CHMPFL12076.POR FLUO012076-25G
3',5'-Dimethyl-2'-hydroxyacetophenon
3',5'-Dimethyl-2'-hydroxyacetophenon
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2000 g/mol
MDL: MFCD01631133
CAS-Nummer: 1198-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR