N-Benzyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(N-Benzylsulphonamido)benzeneboronic acid, (4-(N-Benzylsulphamoyl)phenyl)boronic acid

BB-2419-1G BB-2419-5G
COBBBB-2419-1GEA 210 EUR
COBBBB-2419-1G COBBBB-2419-5G
N-Benzyl-4-boronobenzolsulfonamid
N-Benzyl-4-boronobenzolsulfonamid
Formel: C₁₃H₁₄BNO₄S
Molecular Weight: 291.1306 g/mol
MDL: MFCD06659845
CAS-Nummer: 548769-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR