4-Brom-N-(4-methoxybenzyl)-2-nitroanilin

Lieferant: COMBI-BLOCKS

AN-1267-1G AN-1267-25G AN-1267-5G
COBBAN-1267-1GEA 98.3 EUR
COBBAN-1267-1G COBBAN-1267-25G COBBAN-1267-5G
4-Brom-N-(4-methoxybenzyl)-2-nitroanilin
4-Brom-N-(4-methoxybenzyl)-2-nitroanilin
Formel: C₁₄H₁₃BrN₂O₃
MDL: MFCD09878316
CAS-Nummer: 957062-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR