3-Benzyloxy-2,4-dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Benzyloxy)-2,4-dichlorophenylboronic acid

BB-5220-5G BB-5220-1G
COBBBB-5220-5GEA 1850 EUR
COBBBB-5220-5G COBBBB-5220-1G
3-Benzyloxy-2,4-dichlorphenylboronsäure
3-Benzyloxy-2,4-dichlorphenylboronsäure
Formel: C₁₃H₁₁BCl₂O₃
Molecular Weight: 296,942 g/mol
MDL: MFCD10699639
CAS-Nummer: 1072946-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR