2-Benzyloxy-3-chlorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-(Benzyloxy)-3-chlorophenyl)boronic acid
BB-5213-5G
BB-5213-1G
BB-5213-250M
COBBBB-5213-5GEA
884
EUR
COBBBB-5213-5G
COBBBB-5213-1G
COBBBB-5213-250M
2-Benzyloxy-3-chlorphenylboronsäure
2-Benzyloxy-3-chlorphenylboronsäure
Formel:
C₁₃H₁₂BClO₃ Molecular Weight: 262,5000 g/mol |
MDL:
MFCD13195644 CAS-Nummer: 1217500-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...