2-Benzyloxy-5-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Benzyloxy)-5-chlorophenyl)boronic acid

BB-2606-1G BB-2606-5G
COBBBB-2606-1GEA 166 EUR
COBBBB-2606-1G COBBBB-2606-5G
2-Benzyloxy-5-chlorphenylboronsäure
2-Benzyloxy-5-chlorphenylboronsäure
Formel: C₁₃H₁₂BClO₃
Molecular Weight: 262.4966 g/mol
MDL: MFCD04039000
CAS-Nummer: 612832-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR