3-Benzyloxy-2-chlor-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Benzyoxy-2-chloro-6-fluorophenylboronic acid

BB-5024-5G BB-5024-1G
COBBBB-5024-5GEA 302 EUR
COBBBB-5024-5G COBBBB-5024-1G
3-Benzyloxy-2-chlor-6-fluorphenylboronsäure
3-Benzyloxy-2-chlor-6-fluorphenylboronsäure
Formel: C₁₃H₁₁BClFO₃
Molecular Weight: 280,4900 g/mol
MDL: MFCD09878328
CAS-Nummer: 957062-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR