4-(2-Fluorbenzyloxy)benzaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4-((2-Fluorobenzyl)oxy)benzaldehyde

005934-5G 005934-1G
FLUO005934-5GEA 174 EUR
FLUO005934-5G FLUO005934-1G
4-(2-Fluorbenzyloxy)benzaldehyd
4-(2-Fluorbenzyloxy)benzaldehyd
Formel: C₁₄H₁₁FO₂
MDL: MFCD01590568
CAS-Nummer: 70627-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR