6-Fluor-2-chinolinol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Fluoroquinolin-2(1H)-one

PY-1184-25G PY-1184-1G PY-1184-5G
COBBPY-1184-25GEA 1850 EUR
COBBPY-1184-25G COBBPY-1184-1G COBBPY-1184-5G
6-Fluor-2-chinolinol
6-Fluor-2-chinolinol
Formel: C₉H₆FNO
Molecular Weight: 163,1485 g/mol
MDL: MFCD08236766
CAS-Nummer: 22614-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR