8-Fluor-2-chinolinol

Lieferant: COMBI-BLOCKS

Synonyme: 8-Fluoroquinolin-2(1H)-one

PY-7952-1G PY-7952-5G
COBBPY-7952-1GEA 188 EUR
COBBPY-7952-1G COBBPY-7952-5G
8-Fluor-2-chinolinol
8-Fluor-2-chinolinol
Formel: C₉H₆FNO
Molecular Weight: 163,1500 g/mol
MDL: MFCD12026113
CAS-Nummer: 71738-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR