3-Methoxyisatosäureanhydrid

Lieferant: CHEMPUR

05-0002.1 05-0002.5
CHMP05-0002.1EA 0 EUR
CHMP05-0002.1 CHMP05-0002.5
3-Methoxyisatosäureanhydrid
3-Methoxyisatosäureanhydrid
Formel: C₉H₇NO₄
MDL: MFCD04114326
CAS-Nummer: 34954-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR