4,5-Dimethoxyisatosäureanhydrid

Lieferant: FLUOROCHEM

Synonyme: 6,7-Dimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione

011695-5G 011695-1G
FLUO011695-5GEA 177 EUR
FLUO011695-5G FLUO011695-1G
4,5-Dimethoxyisatosäureanhydrid
4,5-Dimethoxyisatosäureanhydrid
Formel: C₁₀H₉NO₅
Molecular Weight: 223,1800 g/mol
MDL: MFCD03426390
CAS-Nummer: 20197-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR