5-Hydroxyisatosäureanhydrid

Lieferant: FLUOROCHEM

Synonyme: 6-Hydroxy-1H-benzo[d][1,3]oxazine-2,4-dione

011759-10G 011759-5G
FLUO011759-10GEA 0 EUR
FLUO011759-10G FLUO011759-5G
5-Hydroxyisatosäureanhydrid
5-Hydroxyisatosäureanhydrid
Formel: C₈H₅NO₄
Molecular Weight: 179,1296 g/mol
MDL: MFCD00798201
CAS-Nummer: 195986-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR