Aluminiumtitanoxid ≥99,5% (Metall-Basis), Pulver -100 Mesh

Lieferant: Thermo Scientific
Achtung

Synonyme: Aluminum titanate

14484.14 14484.22
14484.14EA 104 EUR
14484.14 14484.22
Aluminiumtitanoxid ≥99,5% (Metall-Basis), Pulver -100 Mesh
Aluminum titanium oxide

Aluminum titanium oxide is used in refractory industry.


Purity: 99.5% CAS No.: 12004-39-6 Molecular Formula: Al2TiO5, may contain small amounts of other phases Formula Weight: 181.86 Physical Form: -100 Mesh Powder Odour: Odourless MDL No.: MFCD00064749

Formel: Al₂O₅Ti
Molecular Weight: 181,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064749
CAS-Nummer: 12004-39-6
EINECS: 234-456-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR