Lanthan(III)isopropoxid (40% La)

Lieferant: Thermo Scientific
Gefahr

14616.02 14616.04 14616.09
14616.02EA 67.1 EUR
14616.02 14616.04 14616.09
Lanthan(III)isopropoxid (40% La)
Lanthan(III)isopropoxid

CAS No.: 19446-52-7 Molecular Formula: C9H21LaO3 Formula Weight: 316.18 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White Odour: Alcohol-like Melting Point: 120-128° Boiling Point: 170°/0.04mm subl. UN No.: 2925 MDL No.: MFCD00015305

Formel: C₉H₂₁LaO₃
Molecular Weight: 316,18 g/mol
Siedepunkt: 170 °C (0,04 mmHg) (Subl.)
Schmelzpunkt: 120…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015305
CAS-Nummer: 19446-52-7
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR