Lanthan(III)isopropoxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

665193-3G 665193-500MG
SIAL665193-3GEA 400 EUR
SIAL665193-3G SIAL665193-500MG
Lanthan(III)isopropoxid, Sigma-Aldrich®
Lanthan(III)isopropoxid
Formel: C₉H₂₁LaO₃
Molecular Weight: 316,18 g/mol
MDL: MFCD00015305
CAS-Nummer: 19446-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR