Ytterbium(III)isopropoxid

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ytterbium(III) i-propoxide

Unter Argon in Ampullen verpackt

39579.03 39579.06
39579.03EA 133 EUR
39579.03 39579.06
Ytterbium(III)isopropoxid
Ytterbium(III)isopropoxid

CAS No.: 6742-69-4 Molecular Formula: C9H21O3Yb Formula Weight: 350.31 Physical Form: Powder, Ampouled under argon Sensitivity: Air and Moisture Sensitive Odour: Odourless Boiling Point: 190-200°/0.2mm subl. UN No.: 2925 MDL No.: MFCD00145532

Formel: C₉H₂₁O₃Yb
Siedepunkt: 190…200 °C (0,2 mmHg) (Subl.)
Schmelzpunkt: 200 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00145532
CAS-Nummer: 6742-69-4
UN: 2925
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR