Tetrahydropyran-4-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Tetrahydropyran-4-yl-carboxylic acid
AM-1006-5G
AM-1006-25G
AM-1006-1G
COBBAM-1006-5GEA
38.5
EUR
COBBAM-1006-5G
COBBAM-1006-25G
COBBAM-1006-1G
Tetrahydropyran-4-carbonsäure
Tetrahydropyran-4-carbonsäure
Formel:
C₆H₁₀O₃ Molecular Weight: 130,14 g/mol |
MDL:
MFCD00031016 CAS-Nummer: 5337-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...