2-Amino-5-brom-4-methoxynicotinonitril

Lieferant: COMBI-BLOCKS

HC-6318-5G HC-6318-1G
COBBHC-6318-5GEA 1150 EUR
COBBHC-6318-5G COBBHC-6318-1G
2-Amino-5-brom-4-methoxynicotinonitril
2-Amino-5-brom-4-methoxynicotinonitril
Formel: C₇H₆BrN₃O
MDL: MFCD09801018
CAS-Nummer: 951884-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR