2-Chlornicotinoylchlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
458821-25G
458821-5G
SIAL458821-25GEA
104
EUR
SIAL458821-25G
SIAL458821-5G
2-Chlornicotinoylchlorid, Sigma-Aldrich®
2-Chlornicotinoylchlorid
Formel:
C₆H₃Cl₂NO Molecular Weight: 176,00 g/mol |
MDL:
MFCD00051677 CAS-Nummer: 49609-84-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...