2-Amino-4-methoxynicotinonitril

Lieferant: BIONET RESEARCH

Synonyme: 2-Amino-4-methoxy-3-pyridinecarbonitrile

2K-027.500 2K-027.1 2K-027.5 2K-027.10
BIOT2K-027.500EA 0 EUR
BIOT2K-027.500 BIOT2K-027.1 BIOT2K-027.5 BIOT2K-027.10
2-Amino-4-methoxynicotinonitril
2-Amino-4-methoxynicotinonitril
Formel: C₇H₇N₃O
Molecular Weight: 149.1500 g/mol
MDL: MFCD00975441
CAS-Nummer: 98651-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR