2-Amino-4-methoxynicotinonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Amino-4-methoxy-3-pyridinecarbonitrile

HC-6193-5G HC-6193-1G
COBBHC-6193-5GEA 662 EUR
COBBHC-6193-5G COBBHC-6193-1G
2-Amino-4-methoxynicotinonitril
2-Amino-4-methoxynicotinonitril
Formel: C₇H₇N₃O
Molecular Weight: 149.1500 g/mol
MDL: MFCD00975441
CAS-Nummer: 98651-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR