2-Amino-3-chlor-5-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

OS-7414-5G OS-7414-25G OS-7414-100G
COBBOS-7414-5GEA 25.5 EUR
COBBOS-7414-5G COBBOS-7414-25G COBBOS-7414-100G
2-Amino-3-chlor-5-(trifluormethyl)pyridin
2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Formel: C₆H₄ClF₃N₂
Molecular Weight: 196,5600 g/mol
MDL: MFCD00042154
CAS-Nummer: 79456-26-1
EINECS: 401-670-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR