6-(Trifluormethyl)nicotinoylchlorid

Lieferant: FLUOROCHEM

009368-1G 009368-5G
FLUO009368-1GEA 96.6 EUR
FLUO009368-1G FLUO009368-5G
6-(Trifluormethyl)nicotinoylchlorid
6-(Trifluormethyl)nicotinoylchlorid
Formel: C₇H₃ClF₃NO
Molecular Weight: 209,5530 g/mol
MDL: MFCD01862645
CAS-Nummer: 358780-13-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR