6-(Trifluormethyl)nicotinoylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

640069-1G 640069-5G
SIAL640069-1GEA 124 EUR
SIAL640069-1G SIAL640069-5G
6-(Trifluormethyl)nicotinoylchlorid, Sigma-Aldrich®
6-(Trifluormethyl)nicotinoylchlorid
Formel: C₇H₃ClF₃NO
Molecular Weight: 209,5530 g/mol
MDL: MFCD01862645
CAS-Nummer: 358780-13-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR