6-(Trifluormethyl)nicotinsäurehydrazid
Lieferant: FLUOROCHEM
Synonyme:
2-(Trifluoromethyl)pyridine-5-carboxylic acid hydrazide
009366-1G
009366-2.5G
FLUO009366-1GEA
186
EUR
FLUO009366-1G
FLUO009366-2.5G
6-(Trifluormethyl)nicotinsäurehydrazid
6-(Trifluormethyl)nicotinsäurehydrazid
Formel:
C₇H₆F₅N₃O |
MDL:
MFCD01862651 CAS-Nummer: 386715-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...