2-Aminophenyl-2-pyridylketon
Lieferant: FLUOROCHEM
011081-1G
011081-5G
FLUO011081-1GEA
294
EUR
FLUO011081-1G
FLUO011081-5G
2-Aminophenyl-2-pyridylketon
2-Aminophenyl-2-pyridylketon
Formel:
C₁₂H₁₀N₂O |
MDL:
MFCD03839744 CAS-Nummer: 42471-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...