2-Amino-4,6-dimethylnicotinamid

Lieferant: BIONET RESEARCH

9C-018.500 9C-018.1 9C-018.5 9C-018.10
BIOT9C-018.500EA 0 EUR
BIOT9C-018.500 BIOT9C-018.1 BIOT9C-018.5 BIOT9C-018.10
2-Amino-4,6-dimethylnicotinamid
2-Amino-4,6-dimethylnicotinamid
Formel: C₈H₁₁N₃O
MDL: MFCD00138572
CAS-Nummer: 7144-20-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR