2-Methyl-6-(trifluormethyl)nicotinsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H33003.03
H33003.09
H33003.03EA
48.2
EUR
H33003.03
H33003.09
2-Methyl-6-(trifluormethyl)nicotinsäure ≥97%
2-Methyl-6-(trifluormethyl)nicotinsäure
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205.14 g/mol Schmelzpunkt: 139…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311987 CAS-Nummer: 261635-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...