2-Methyl-6-(trifluormethyl)nicotinsäure

Lieferant: Thermo Scientific

AW00680DA AW00680EA
MAYBAW00680DAEA 0 EUR
MAYBAW00680DA MAYBAW00680EA
2-Methyl-6-(trifluormethyl)nicotinsäure
2-Methyl-6-(trifluormethyl)nicotinsäure
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD01311987
CAS-Nummer: 261635-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR