6-(4-Fluorphenyl)-2-pyridincarbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-(4-Fluorophenyl)picolinaldehyde, 6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

654477-1G 654477-5G
SIAL654477-1GEA 82.5 EUR
SIAL654477-1G SIAL654477-5G
6-(4-Fluorphenyl)-2-pyridincarbaldehyd, Sigma-Aldrich®
6-(4-Fluorphenyl)-2-pyridincarbaldehyd
Formel: C₁₂H₈FNO
Molecular Weight: 201,196 g/mol
MDL: MFCD06410011
CAS-Nummer: 834884-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR