2-Thiophenboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(2-thienyl)-1,3,2-dioxaborolane

PN-2339-5G PN-2339-25G PN-2339-1G
COBBPN-2339-5GEA 45.9 EUR
COBBPN-2339-5G COBBPN-2339-25G COBBPN-2339-1G
2-Thiophenboronsäurepinakolester
2-Thiophenboronsäurepinakolester
Formel: C₁₀H₁₅BO₂S
Molecular Weight: 210.1 g/mol
MDL: MFCD05663878
CAS-Nummer: 193978-23-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR