3-Thiophenboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Thiophene-3-boronic acid pinacol ester, Thiophene-3-boronic acid, pinacol ester

PN-2553-5G PN-2553-25G PN-2553-1G
COBBPN-2553-5GEA 243 EUR
COBBPN-2553-5G COBBPN-2553-25G COBBPN-2553-1G
3-Thiophenboronsäurepinakolester
3-Thiophenboronsäurepinakolester
Formel: C₁₀H₁₅BO₂S
Molecular Weight: 210,1009 g/mol
MDL: MFCD05663893
CAS-Nummer: 214360-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR