2-Brombenzaldehydethylenacetal, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(2-Bromophenyl)-1,3-dioxolane
M-Clarity™ Qualitätsstufe = MQ100
652652-25G
652652-5G
SIAL652652-25GEA
192
EUR
SIAL652652-25G
SIAL652652-5G
2-Brombenzaldehydethylenacetal, Sigma-Aldrich®
2-Brombenzaldehydethylenacetal
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol |
MDL:
MFCD00155124 CAS-Nummer: 34824-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...