Tris(pentafluorphenyl)boran

Lieferant: FLUOROCHEM

Synonyme: Tris(perfluorophenyl)borane

010564-5G 010564-1G
FLUO010564-5GEA 0 EUR
FLUO010564-5G FLUO010564-1G
Tris(pentafluorphenyl)boran
Tris(pentafluorphenyl)boran
Formel: C₁₈BF₁₅
Molecular Weight: 511,98 g/mol
MDL: MFCD00269813
CAS-Nummer: 1109-15-5
EINECS: 213-504-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR