Maltohexaose, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal

M-Clarity™ Qualitätsstufe = MQ200

M9153-100MG M9153-500MG
SIALM9153-100MGEA 261 EUR
SIALM9153-100MG SIALM9153-500MG
Maltohexaose, Sigma-Aldrich®
Maltohexaose
Formel: C₃₆H₆₂O₃₁
Molecular Weight: 990,8589 g/mol
MDL: MFCD00083650
CAS-Nummer: 34620-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR