Di(2-pyridyl)keton, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Di(pyridin-2-yl)methanone
M-Clarity™ Qualitätsstufe = MQ100
127221-25G
127221-5G
SIAL127221-25GEA
607
EUR
SIAL127221-25G
SIAL127221-5G
Di(2-pyridyl)keton, Sigma-Aldrich®
Di(2-pyridyl)keton
Formel:
C₁₁H₈N₂O Molecular Weight: 184,20 g/mol |
MDL:
MFCD00006288 CAS-Nummer: 19437-26-4 EINECS: 243-067-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...