10,11-Dihydrocarbamazepin ≥99%

Lieferant: Thermo Scientific

Synonyme: 10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carboxamide

L17271.MD L17271.MF
L17271.MDEA 122 EUR
L17271.MD L17271.MF
10,11-Dihydrocarbamazepin ≥99%
10,11-Dihydrocarbamazepin

Purity: 99% CAS No.: 3564-73-6 Molecular Formula: C15H14N2O Formula Weight: 238.29 Melting Point: 204-206° Beilstein Reference: 196969

Formel: C₁₅H₁₄N₂O
Molecular Weight: 238,29 g/mol
Schmelzpunkt: 204…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005072
CAS-Nummer: 3564-73-6
EINECS: 222-649-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR