1-(2-Tetrahydropyranyl)-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole

694665-1G 694665-5G
SIAL694665-1GEA 64.2 EUR
SIAL694665-1G SIAL694665-5G
1-(2-Tetrahydropyranyl)-1H-pyrazol, Sigma-Aldrich®
1-(2-Tetrahydropyranyl)-1H-pyrazol
Formel: C₈H₁₂N₂O
Molecular Weight: 152,20 g/mol
MDL: MFCD11044692
CAS-Nummer: 449758-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR