2,3-Dibrom-3-phenylpropiophenon

Lieferant: BIONET RESEARCH

7G-914.500 7G-914.1
BIOT7G-914.500EA 0 EUR
BIOT7G-914.500 BIOT7G-914.1
2,3-Dibrom-3-phenylpropiophenon
2,3-Dibrom-3-phenylpropiophenon
Formel: C₁₅H₁₂Br₂O
MDL: MFCD00017861
CAS-Nummer: 611-91-6
EINECS: 210-282-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR