6-Bromchinolin-2(1H)-on 96%

Lieferant: Thermo Scientific
Achtung

H54095.03 H54095.06
H54095.03EA 60.8 EUR
H54095.03 H54095.06
6-Bromchinolin-2(1H)-on 96%
6-Bromchinolin-2(1H)-on

Purity: 96% CAS No.: 1810-66-8 Molecular Formula: C9H6BrNO Formula Weight: 224.05 Storage Temperature: 2°C to 8°C Melting Point: 200° MDL No.: MFCD08703169

Formel: C₉H₆BrNO
Molecular Weight: 224,05 g/mol
Schmelzpunkt: >200 °C
MDL: MFCD08703169
CAS-Nummer: 1810-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR